siRNA Details
| Gene Symbol: Slc22a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031332.1 | NP_112622.1 | 11 | 1667 | ||
| GenBank mRNA | AB017446.1 | - | 11 | 1667 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a8| Gene Symbol: Slc22a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031332.1 | NP_112622.1 | 11 | 1667 | ||
| GenBank mRNA | AB017446.1 | - | 11 | 1667 | ||