siRNA Details
| Gene Symbol: St3gal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031337.2 | NP_112627.2 | 4, 5 | 799 | ||
| XM_006236676.1 | XP_006236738.1 | 3, 4 | 460 | |||
| XM_006236677.1 | XP_006236739.1 | 4, 5 | 602 | |||
| XM_006236679.2 | XP_006236741.1 | 3, 4 | 593 | |||
| XM_006236680.1 | XP_006236742.1 | 3, 4 | 460 | |||
| XM_008762996.1 | XP_008761218.1 | 3, 4 | 622 | |||
| GenBank mRNA | AB018049.1 | - | 3, 4 | 465 | ||
| BC078798.1 | - | 4, 5 | 799 | 7106217 | ||

St3gal5
Genomic Map