siRNA Details
| Gene Symbol: Camkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031338.1 | NP_112628.1 | 2 | 265 | ||
| GenBank mRNA | AB018081.1 | - | 2 | 265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Camkk2| Gene Symbol: Camkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031338.1 | NP_112628.1 | 2 | 265 | ||
| GenBank mRNA | AB018081.1 | - | 2 | 265 | ||