siRNA Details
| Gene Symbol: Ppargc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031347.1 | NP_112637.1 | 3, 4 | 463 | ||
| XM_008770219.1 | XP_008768441.1 | 3, 4 | 530 | |||
| XM_008770220.1 | XP_008768442.1 | 2, 3 | 1700 | |||
| GenBank mRNA | AB025784.1 | - | 3, 4 | 463 | ||
| AY237127.1 | - | 3, 4 | 422 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008904, AF049330, AB061324, AK138668, BC066868, and others.

Ppargc1a
Genomic Map