siRNA Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031348.2 | NP_112638.2 | 9 | 762 | ||
| XM_008761614.1 | XP_008759836.1 | 9 | 785 | |||
| GenBank mRNA | AB026057.2 | - | 9 | 673 | ||
| BC088128.1 | - | 9 | 762 | 7376326 | ||

Fcna
Genomic Map