siRNA Details
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031561.2 | NP_113749.2 | 334 | |||
| GenBank mRNA | AB005743.1 | - | 396 | |||
| AF072411.1 | - | 1 | 125 | |||
| AF111268.1 | - | 141 | ||||
| AF113914.1 | - | 1 | 119 | |||
| BC072543.1 | - | 334 | 7105943 | |||
| L19658.1 | - | 1 | 174 | |||

Cd36
Genomic Map