siRNA Details
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031561.2 | NP_113749.2 | 895 | |||
| GenBank mRNA | AF072411.1 | - | 5 | 686 | ||
| AF113914.1 | - | 5 | 680 | |||
| BC072543.1 | - | 895 | 7105943 | |||
| L19658.1 | - | 5 | 735 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd36| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031561.2 | NP_113749.2 | 895 | |||
| GenBank mRNA | AF072411.1 | - | 5 | 686 | ||
| AF113914.1 | - | 5 | 680 | |||
| BC072543.1 | - | 895 | 7105943 | |||
| L19658.1 | - | 5 | 735 | |||