siRNA Details
| Gene Symbol: Wisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031590.1 | NP_113778.1 | 5 | 929 | ||
| GenBank mRNA | AF259981.1 | - | 5 | 929 | ||
| BC089187.1 | - | 4 | 739 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wisp2| Gene Symbol: Wisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031590.1 | NP_113778.1 | 5 | 929 | ||
| GenBank mRNA | AF259981.1 | - | 5 | 929 | ||
| BC089187.1 | - | 4 | 739 | |||