siRNA Details
| Gene Symbol: Rabggta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031654.2 | NP_113842.1 | 3 | 170 | ||
| XM_006252009.2 | XP_006252071.1 | 3 | 265 | |||
| XM_006252010.2 | XP_006252072.1 | 3 | 212 | |||
| GenBank mRNA | BC086547.1 | - | 3 | 170 | 7301723 | |
| L10415.1 | - | 130 | ||||
| S62096.1 | - | 130 | ||||

Rabggta
Genomic Map