siRNA Details
| Gene Symbol: Mmp24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031757.1 | NP_113945.1 | 4, 5 | 534 | ||
| GenBank mRNA | AB023659.1 | - | 4, 5 | 534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp24| Gene Symbol: Mmp24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031757.1 | NP_113945.1 | 4, 5 | 534 | ||
| GenBank mRNA | AB023659.1 | - | 4, 5 | 534 | ||