siRNA Details
| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031769.1 | NP_113957.1 | 4 | 314 | ||
| GenBank mRNA | AF020346.1 | - | 4 | 314 | ||
| FQ213832.1 | - | 4 | 418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdxk| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031769.1 | NP_113957.1 | 4 | 314 | ||
| GenBank mRNA | AF020346.1 | - | 4 | 314 | ||
| FQ213832.1 | - | 4 | 418 | |||