siRNA Details
| Gene Symbol: Gda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031776.2 | NP_113964.2 | 2 | 204 | ||
| GenBank mRNA | AF026472.2 | - | 2 | 261 | ||
| AF245172.1 | - | 2 | 204 | |||
| FQ233257.1 | - | 2 | 226 | |||
| FQ233523.1 | - | 2 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gda| Gene Symbol: Gda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031776.2 | NP_113964.2 | 2 | 204 | ||
| GenBank mRNA | AF026472.2 | - | 2 | 261 | ||
| AF245172.1 | - | 2 | 204 | |||
| FQ233257.1 | - | 2 | 226 | |||
| FQ233523.1 | - | 2 | 232 | |||