siRNA Details
Gene Symbol: Dedd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031800.1 | NP_113988.1 | 1 | 319 | ||
XM_006250269.2 | XP_006250331.1 | 3 | 384 | |||
XM_006250270.2 | XP_006250332.1 | 1 | 334 | |||
XM_006250272.2 | XP_006250334.1 | 3 | 464 | |||
XM_006250273.2 | XP_006250335.1 | 2 | 390 | |||
XM_006250274.2 | XP_006250336.1 | 1 | 338 | |||
GenBank mRNA | AF053362.1 | - | 1 | 319 |
