siRNA Details
| Gene Symbol: Kcnma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031828.1 | NP_114016.1 | 16, 18 | 2434 | ||
| GenBank mRNA | AF083341.1 | - | 16, 18 | 1870 | ||
| AF135265.1 | - | 16, 18 | 2288 | |||
| U40603.1 | - | 16, 18 | 2434 | |||

Kcnma1
Genomic Map