siRNA Details
| Gene Symbol: Src | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031977.1 | NP_114183.1 | 12 | 1631 | ||
| XM_008762387.1 | XP_008760609.1 | 12 | 1640 | |||
| XM_008762388.1 | XP_008760610.1 | 12 | 1677 | |||
| XM_008762389.1 | XP_008760611.1 | 12 | 1748 | |||
| GenBank mRNA | AF157016.1 | - | 12 | 1631 | ||

Src
Genomic Map