siRNA Details
| Gene Symbol: Sdcbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031986.1 | NP_114192.1 | 9 | 1482 | ||
| GenBank mRNA | AF248548.1 | - | 9 | 1482 | ||
| AJ292243.1 | - | 8 | 1428 | |||
| BC064651.1 | - | 9 | 1453 | 5598152 | ||
| FQ225374.1 | - | 1 | 526 | |||
| FQ230274.1 | - | 1 | 526 | |||

Sdcbp
Genomic Map