siRNA Details
| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032073.1 | NP_114462.1 | 3, 4 | 685 | ||
| GenBank mRNA | AJ133685.1 | - | 3, 4 | 685 | ||
| AY043394.1 | - | 2, 3 | 425 | |||
| U92655.1 | - | 2, 3 | 197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnq1| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032073.1 | NP_114462.1 | 3, 4 | 685 | ||
| GenBank mRNA | AJ133685.1 | - | 3, 4 | 685 | ||
| AY043394.1 | - | 2, 3 | 425 | |||
| U92655.1 | - | 2, 3 | 197 | |||