siRNA Details
| Gene Symbol: Akt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033230.2 | NP_150233.1 | 2 | 147 | ||
| XM_006240631.2 | XP_006240693.1 | 3 | 646 | |||
| XM_008764918.1 | XP_008763140.1 | 2 | 2193 | |||
| GenBank mRNA | D30040.1 | - | 2 | 151 | ||

Akt1
Genomic Map