siRNA Details
| Gene Symbol: Ngb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033359.3 | NP_203523.2 | 4 | 1465 | ||
| XM_008764864.1 | XP_008763086.1 | 4 | 2158 | |||
| GenBank mRNA | AY066001.1 | - | 4 | 1250 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ngb| Gene Symbol: Ngb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033359.3 | NP_203523.2 | 4 | 1465 | ||
| XM_008764864.1 | XP_008763086.1 | 4 | 2158 | |||
| GenBank mRNA | AY066001.1 | - | 4 | 1250 | ||