siRNA Details
| Gene Symbol: Klf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053394.2 | NP_445846.2 | 2 | 1379 | ||
| XM_006252403.2 | XP_006252465.1 | 2 | 1016 | |||
| GenBank mRNA | AF182101.1 | - | 1 | 533 | ||
| BC081758.1 | - | 2 | 1021 | 7132572 | ||

Klf5
Genomic Map