siRNA Details
| Gene Symbol: Pomt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053406.2 | NP_445858.1 | 4, 5 | 439 | ||
| XM_006234001.2 | XP_006234063.1 | 4, 5 | 1041 | |||
| XM_008761776.1 | XP_008759998.1 | 2, 3 | 149 | |||
| GenBank mRNA | AF192388.1 | - | 4, 5 | 412 | ||
| BC070912.1 | - | 4, 5 | 409 | 7104668 | ||
| DY309994.1 | - | 4, 5 | 439 | |||

Pomt1
Genomic Map