siRNA Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053416.2 | NP_445868.2 | 3 | 548 | ||
| XM_006234108.2 | XP_006234170.2 | 4 | 886 | |||
| XM_008761777.1 | XP_008759999.1 | 4 | 586 | |||
| XM_008761778.1 | XP_008760000.1 | 4 | 586 | |||
| GenBank mRNA | CO564209.1 | - | 4 | 625 | ||

Strbp
Genomic Map