siRNA Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053418.2 | NP_445870.1 | 5, 6 | 521 | ||
| XM_008768904.1 | XP_008767126.1 | 5, 6 | 538 | |||
| GenBank mRNA | AF234601.1 | - | 5, 6 | 496 | ||
| BC161818.1 | - | 5, 6 | 532 | 9101868 | ||
| CK599711.1 | - | 521 | ||||

Ufd1l
Genomic Map