siRNA Details
| Gene Symbol: Ccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053425.1 | NP_445877.1 | 5 | 506 | ||
| GenBank mRNA | AF255305.1 | - | 5 | 506 | ||
| BC158586.1 | - | 5 | 466 | 9099085 | ||
| EU340033.1 | - | 4 | 368 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_016892, AK192205, AF121906, AK010264, AK050072, and others.

Ccs
Genomic Map