siRNA Details
| Gene Symbol: Fgf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053428.1 | NP_445880.1 | 2, 3 | 256 | ||
| XM_006257592.1 | XP_006257654.1 | 2, 3 | 524 | |||
| XM_008773572.1 | XP_008771794.1 | 3, 4 | 432 | |||
| XM_008773573.1 | XP_008771795.1 | 3, 4 | 494 | |||
| GenBank mRNA | AF271786.1 | - | 2, 3 | 256 | ||

Fgf13
Genomic Map