siRNA Details
| Gene Symbol: Dgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053437.1 | NP_445889.1 | 7 | 735 | ||
| GenBank mRNA | AB062759.1 | - | 7 | 713 | ||
| AB062760.1 | - | 7 | 713 | |||
| AB062761.1 | - | 7 | 713 | |||
| AB062762.1 | - | 7 | 713 | |||
| AB062763.1 | - | 7 | 711 | |||
| AF296131.1 | - | 7 | 735 | |||
| AJ345014.1 | - | 1 | 23 | |||
| BC081742.1 | - | 7 | 707 | |||

Dgat1
Genomic Map