siRNA Details
| Gene Symbol: Rab9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053458.1 | NP_445910.1 | 3 | 534 | ||
| GenBank mRNA | AF325692.1 | - | 3 | 534 | ||
| BC070502.1 | - | 3 | 538 | 7096737 | ||
| FQ212962.1 | - | 3 | 543 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab9a| Gene Symbol: Rab9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053458.1 | NP_445910.1 | 3 | 534 | ||
| GenBank mRNA | AF325692.1 | - | 3 | 534 | ||
| BC070502.1 | - | 3 | 538 | 7096737 | ||
| FQ212962.1 | - | 3 | 543 | |||