siRNA Details
Gene Symbol: Ptp4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053475.1 | NP_445927.1 | 1 | 484 | ||
GenBank mRNA | AJ007016.1 | - | 1 | 484 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptp4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053475.1 | NP_445927.1 | 1 | 484 | ||
GenBank mRNA | AJ007016.1 | - | 1 | 484 |