siRNA Details
| Gene Symbol: Mlycd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053477.1 | NP_445929.1 | 2 | 542 | ||
| GenBank mRNA | AF304865.1 | - | 2 | 542 | ||
| AJ007704.1 | - | 2 | 542 | |||
| BC061845.1 | - | 2 | 545 | 5621015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlycd| Gene Symbol: Mlycd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053477.1 | NP_445929.1 | 2 | 542 | ||
| GenBank mRNA | AF304865.1 | - | 2 | 542 | ||
| AJ007704.1 | - | 2 | 542 | |||
| BC061845.1 | - | 2 | 545 | 5621015 | ||