siRNA Details
| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243642.2 | XP_006243704.1 | 2 | 208 | ||
| GenBank mRNA | AJ012482.1 | - | 2 | 191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pik3cb| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243642.2 | XP_006243704.1 | 2 | 208 | ||
| GenBank mRNA | AJ012482.1 | - | 2 | 191 | ||