siRNA Details
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053484.2 | NP_445936.2 | 6 | 425 | ||
| XM_006246827.2 | XP_006246889.1 | 6 | 1094 | |||
| XM_006246828.2 | XP_006246890.1 | 6 | 1935 | |||
| XM_006246829.2 | XP_006246891.1 | 6 | 1278 | |||
| XM_006246830.2 | XP_006246892.1 | 6 | 609 | |||
| XM_008767883.1 | XP_008766105.1 | 6 | 1009 | |||
| GenBank mRNA | AJ003148.1 | - | 6 | 561 | ||
| AJ131902.1 | - | 561 | ||||

Gas7
Genomic Map