siRNA Details
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053484.2 | NP_445936.2 | 14 | 3412 | ||
| XM_006246827.2 | XP_006246889.1 | 14 | 4081 | |||
| XM_006246828.2 | XP_006246890.1 | 14 | 4922 | |||
| XM_006246829.2 | XP_006246891.1 | 14 | 4265 | |||
| XM_006246830.2 | XP_006246892.1 | 14 | 3596 | |||
| XM_008767883.1 | XP_008766105.1 | 14 | 3996 | |||
| GenBank mRNA | AJ131902.1 | - | 3554 | |||

Gas7
Genomic Map