siRNA Details
| Gene Symbol: Kif3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053486.1 | NP_445938.1 | 1 | 302 | ||
| GenBank mRNA | AF083330.1 | - | 1 | 269 | ||
| AJ223599.1 | - | 1 | 302 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif3c| Gene Symbol: Kif3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053486.1 | NP_445938.1 | 1 | 302 | ||
| GenBank mRNA | AF083330.1 | - | 1 | 269 | ||
| AJ223599.1 | - | 1 | 302 | |||