siRNA Details
| Gene Symbol: Xpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053490.1 | NP_445942.1 | 3, 4 | 295 | ||
| XM_006251578.2 | XP_006251640.1 | 4, 5 | 1091 | |||
| XM_008770437.1 | XP_008768659.1 | 2, 3 | 619 | |||
| GenBank mRNA | AB105193.1 | - | 3, 4 | 295 | ||

Xpo1
Genomic Map