siRNA Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053502.1 | NP_445954.1 | 15 | 2847 | ||
| XM_006256233.2 | XP_006256295.1 | 14 | 3020 | |||
| GenBank mRNA | AJ303374.1 | - | 15 | 2847 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcg1| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053502.1 | NP_445954.1 | 15 | 2847 | ||
| XM_006256233.2 | XP_006256295.1 | 14 | 3020 | |||
| GenBank mRNA | AJ303374.1 | - | 15 | 2847 | ||