siRNA Details
| Gene Symbol: Nol3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053516.2 | NP_445968.2 | 2 | 742 | ||
| XM_006255539.2 | XP_006255601.1 | 3 | 873 | |||
| XM_008772540.1 | XP_008770762.1 | 3 | 1354 | |||
| GenBank mRNA | U40627.1 | - | 2 | 742 | ||

Nol3
Genomic Map