siRNA Details
| Gene Symbol: Herpud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053523.1 | NP_445975.1 | 2, 3 | 324 | ||
| GenBank mRNA | AB033771.1 | - | 2, 3 | 324 | ||
| BC061849.1 | - | 2, 3 | 346 | 5621877 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Herpud1| Gene Symbol: Herpud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053523.1 | NP_445975.1 | 2, 3 | 324 | ||
| GenBank mRNA | AB033771.1 | - | 2, 3 | 324 | ||
| BC061849.1 | - | 2, 3 | 346 | 5621877 | ||