siRNA Details
| Gene Symbol: Herpud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053523.1 | NP_445975.1 | 8 | 1396 | ||
| XM_006255133.2 | XP_006255195.1 | 8 | 1709 | |||
| GenBank mRNA | AB033771.1 | - | 8 | 1396 | ||
| BC061849.1 | - | 8 | 1418 | 5621877 | ||
| FQ219047.1 | - | 3 | 541 | |||

Herpud1
Genomic Map