siRNA Details
| Gene Symbol: Sfrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053544.1 | NP_445996.1 | 6 | 1272 | ||
| GenBank mRNA | AF012891.1 | - | 6 | 1272 | ||
| AF140346.1 | - | 6 | 1423 | |||
| FQ220237.1 | - | 3 | 581 | |||
| FQ229078.1 | - | 3 | 594 | |||
| FQ229224.1 | - | 5 | 703 | |||
More Information
Set Membership: |
|

Sfrp4
Genomic Map