siRNA Details
Gene Symbol: Opn1mw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053548.1 | NP_446000.1 | 2 | 222 | ||
GenBank mRNA | AF031528.1 | - | 1 | 96 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
