siRNA Details
Gene Symbol: Syngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053553.1 | NP_446005.1 | 4 | 1090 | ||
GenBank mRNA | AF039085.1 | - | 4 | 1090 | ||
FQ218897.1 | - | 935 | ||||
FQ224907.1 | - | 935 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Syngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053553.1 | NP_446005.1 | 4 | 1090 | ||
GenBank mRNA | AF039085.1 | - | 4 | 1090 | ||
FQ218897.1 | - | 935 | ||||
FQ224907.1 | - | 935 |