siRNA Details
| Gene Symbol: Psmb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053590.1 | NP_446042.1 | 3 | 295 | ||
| XM_006227981.1 | XP_006228043.1 | 5 | 550 | |||
| GenBank mRNA | BC058455.1 | - | 3 | 307 | 6922317 | |
| FQ211516.1 | - | 3 | 309 | |||
| FQ214442.1 | - | 3 | 310 | |||
| FQ214726.1 | - | 5 | 550 | |||
| FQ218426.1 | - | 3 | 280 | |||
| FQ225328.1 | - | 3 | 310 | |||
| FQ229756.1 | - | 3 | 310 | |||
| FQ230094.1 | - | 3 | 310 | |||
| X52783.1 | - | 3 | 295 | |||

Psmb1
Genomic Map