siRNA Details
Gene Symbol: Pgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053595.2 | NP_446047.1 | 3 | 495 | ||
GenBank mRNA | BC087006.1 | - | 3 | 495 | 7193034 | |
L40030.1 | - | 3 | 230 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053595.2 | NP_446047.1 | 3 | 495 | ||
GenBank mRNA | BC087006.1 | - | 3 | 495 | 7193034 | |
L40030.1 | - | 3 | 230 |