siRNA Details
| Gene Symbol: Clic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053603.2 | NP_446055.1 | 7 | 5598 | ||
| GenBank mRNA | AF323174.1 | - | 7 | 1892 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clic5| Gene Symbol: Clic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053603.2 | NP_446055.1 | 7 | 5598 | ||
| GenBank mRNA | AF323174.1 | - | 7 | 1892 | ||