siRNA Details
| Gene Symbol: Dao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053626.1 | NP_446078.1 | 3 | 305 | ||
| GenBank mRNA | AB003400.1 | - | 3 | 305 | ||
| BC088395.1 | - | 2 | 220 | 7110523 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dao| Gene Symbol: Dao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053626.1 | NP_446078.1 | 3 | 305 | ||
| GenBank mRNA | AB003400.1 | - | 3 | 305 | ||
| BC088395.1 | - | 2 | 220 | 7110523 | ||