siRNA Details
| Gene Symbol: Egr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053633.1 | NP_446085.1 | 2 | 1777 | ||
| XM_008772857.1 | XP_008771079.1 | 3 | 1454 | |||
| GenBank mRNA | AB032419.1 | - | 1 | 1826 | ||
| AB032420.1 | - | 2 | 1777 | |||
| D83508.1 | - | 2 | 1688 | |||

Egr2
Genomic Map