siRNA Details
| Gene Symbol: Pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053650.1 | NP_446102.1 | 2, 3 | 334 | ||
| XM_006253119.2 | XP_006253181.1 | 2, 3 | 369 | |||
| XM_006253120.2 | XP_006253182.1 | 2, 3 | 367 | |||
| GenBank mRNA | AF002281.1 | - | 2, 3 | 352 | ||
| BC081703.1 | - | 2, 3 | 326 | 7193873 | ||
| FQ215471.1 | - | 2, 3 | 349 | |||

Pdlim3
Genomic Map