siRNA Details
| Gene Symbol: Pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053650.1 | NP_446102.1 | 8 | 1292 | ||
| XM_006253119.2 | XP_006253181.1 | 7 | 1191 | |||
| XM_006253120.2 | XP_006253182.1 | 7 | 1069 | |||
| GenBank mRNA | AF002281.1 | - | 8 | 1310 | ||
| BC081703.1 | - | 7 | 1148 | 7193873 | ||
| FQ215471.1 | - | 8 | 1315 | |||

Pdlim3
Genomic Map