siRNA Details
| Gene Symbol: Vegfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053653.1 | NP_446105.1 | 3 | 522 | ||
| GenBank mRNA | AY032729.1 | - | 3 | 522 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vegfc| Gene Symbol: Vegfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053653.1 | NP_446105.1 | 3 | 522 | ||
| GenBank mRNA | AY032729.1 | - | 3 | 522 | ||