siRNA Details
| Gene Symbol: Ccnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053662.3 | NP_446114.1 | 928 | |||
| XM_008761029.1 | XP_008759251.1 | 2004 | ||||
| GenBank mRNA | AF030091.3 | - | 928 | |||
| BC085686.1 | - | 982 | ||||
| BC128699.1 | - | 1022 | ||||
| BC161817.1 | - | 874 | 9022416 | |||
| FQ213168.1 | - | 4 | 352 | |||
| FQ232436.1 | - | 965 | ||||

Ccnl1
Genomic Map